accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 351003-19-5

H3COONN

CAS RN 351003-19-5

IUPAC Name: 3-(6-methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazin-4-yl)propanenitrile
Synonyms: 3-(6-methyl-3-oxo-2H-1,4-benzoxazin-4-yl)propanenitrile
Molecular Weight (g/mol): 216.24
Molecular Formula: C12H12N2O2
InChi Key: SZEGFLHUNYLAOJ-UHFFFAOYSA-N
SMILES: CC1=CC2=C(OCC(=O)N2CCC#N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results