accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 35138-23-9

IUPAC Name: bis(cycloocta-1,5-diene) tetrafluoroboranuide iridium
Synonyms: bis(1,5-cyclooctadiene) iridium tetrafluoroborate
Molecular Weight (g/mol): 495.39
Molecular Formula: C16H24BF4Ir
InChi Key: ZFVHFEXIVQSRNV-UHFFFAOYSA-N
SMILES: [Ir].F[B-](F)(F)F.C1CC=CCCC=C1.C1CC=CCCC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results