accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 35166-37-1

IUPAC Name: 3-(chloromethyl)-5-methyl-1,2-oxazole
Synonyms: 3-(chloromethyl)-5-methyl-1,2-oxazole
Molecular Weight (g/mol): 131.56
Molecular Formula: C5H6ClNO
InChi Key: FEXTXBAFBURKGS-UHFFFAOYSA-N
SMILES: CC1=CC(CCl)=NO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results