accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 35192-73-5

H3COHCH2

CAS RN 35192-73-5

IUPAC Name: non-1-en-4-ol
Synonyms: non-1-en-4-ol
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: NDUPBTSXOSZHON-UHFFFAOYNA-N
SMILES: CCCCCC(O)CC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results