accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FMgBr

CAS RN 352-13-6

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 199.31
Molecular Formula: C6H4BrFMg
InChi Key: —
SMILES: [Mg++].[Br-].Fc1cc[cH-]cc1
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (2)
  • (2)