accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 352018-90-7

IUPAC Name: ethyl 1-(4-cyanophenyl)piperidine-4-carboxylate
Synonyms: ethyl 1-(4-cyanophenyl)piperidine-4-carboxylate
Molecular Weight (g/mol): 258.32
Molecular Formula: C15H18N2O2
InChi Key: LIWHXOHASZYDEM-UHFFFAOYSA-N
SMILES: CCOC(=O)C1CCN(CC1)C1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results