accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHCH2

CAS RN 3521-91-3

IUPAC Name: hept-1-en-4-ol
Synonyms: 1-hepten-4-ol
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: AQTUHJABKZECGA-UHFFFAOYNA-N
SMILES: CCCC(O)CC=C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results