accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOBr

CAS RN 35223-80-4

IUPAC Name: propyl 2-bromoacetate
Synonyms: propyl 2-bromoacetate
Molecular Weight (g/mol): 181.03
Molecular Formula: C5H9BrO2
InChi Key: ISYUCUGTDNJIHV-UHFFFAOYSA-N
SMILES: CCCOC(=O)CBr
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results