accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 35272-15-2

IUPAC Name: 2-methyl-1,3-thiazole-4-carboxylic acid
Synonyms: 2-methyl-1,3-thiazole-4-carboxylic acid
Molecular Weight (g/mol): 143.16
Molecular Formula: C5H5NO2S
InChi Key: ZHDRDZMTEOIWSX-UHFFFAOYSA-N
SMILES: CC1=NC(=CS1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results