accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3NNH2N

CAS RN 353-07-1

IUPAC Name: 1-(2-cyanoethyl)hydrazine-1,2-diium
Synonyms: 1-(2-cyanoethyl)hydrazine-1,2-diium
Molecular Weight (g/mol): 87.13
Molecular Formula: C3H9N3
InChi Key: PDUFDGPPXZKHPU-UHFFFAOYSA-N
SMILES: [NH3+][NH2+]CCC#N
Boiling Point 
  • (5)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results