accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 353-09-3

IUPAC Name: 3-[(diaminomethylidene)amino]propanoic acid
Synonyms: amidino β-alanine
Molecular Weight (g/mol): 131.14
Molecular Formula: C4H9N3O2
InChi Key: KMXXSJLYVJEBHI-UHFFFAOYSA-N
SMILES: NC(N)=NCCC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results