accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SBH3CH3CFFF

CAS RN 353-43-5

IUPAC Name: (dimethyl-λ⁴-sulfanylidene)trifluoro-λ⁵-borane
Synonyms: (dimethyl-λ⁴-sulfanylidene)trifluoro-λ⁵-borane
Molecular Weight (g/mol): 129.94
Molecular Formula: C2H6BF3S
InChi Key: VPECYQXBNOAJIF-UHFFFAOYSA-N
SMILES: C[S+](C)[B-](F)(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results