accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 35305-74-9

H3CONH(S)NH2OOHO

CAS RN 35305-74-9

IUPAC Name: (2S)-4-carbamoyl-2-acetamidobutanoic acid
Synonyms: N-acetyl-L-glutamine
Molecular Weight (g/mol): 188.18
Molecular Formula: C7H12N2O4
InChi Key: KSMRODHGGIIXDV-YFKPBYRVSA-N
SMILES: CC(=O)N[C@@H](CCC(N)=O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results