accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 35370-80-0

H2NSO

CAS RN 35370-80-0

IUPAC Name: 2-phenoxyethanethioamide
Synonyms: 2-phenoxyethanethioamide
Molecular Weight (g/mol): 167.23
Molecular Formula: C8H9NOS
InChi Key: XORPVBXCJIPCBQ-UHFFFAOYSA-N
SMILES: NC(=S)COC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results