accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOOH2NClFFF

CAS RN 35375-74-7

IUPAC Name: 5-chloro-2-nitro-4-(trifluoromethyl)aniline
Synonyms: 5-chloro-2-nitro-4-(trifluoromethyl)aniline
Molecular Weight (g/mol): 240.57
Molecular Formula: C7H4ClF3N2O2
InChi Key: AAMCWLIPRMIPBN-UHFFFAOYSA-N
SMILES: NC1=C(C=C(C(Cl)=C1)C(F)(F)F)[N+]([O-])=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)