accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 35387-93-0

IUPAC Name: methyl 3-iodo-4-methoxybenzoate
Synonyms: methyl 3-iodo-4-methoxybenzoate
Molecular Weight (g/mol): 292.07
Molecular Formula: C9H9IO3
InChi Key: GHNGBFHLUOJHKP-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(OC)C(I)=C1
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results