CAS RN 354-26-7
IUPAC Name:
1-chloro-1,1,2-trifluoro-2-iodoethane
Synonyms:
1-chloro-1,1,2-trifluoro-2-iodoethane
Molecular Weight (g/mol):
244.38
Molecular Formula:
C2HClF3I
InChi Key:
XONGRFUEHPBOBB-UHFFFAOYNA-N
SMILES:
FC(I)C(F)(F)Cl
1
–
1
of
1
results
1
Alfa Aesar™ 1-Chloro-1,1,2-trifluoro-2-iodoethane, 97%
CAS: 354-26-7 Molecular Formula: C2HClF3I Molecular Weight (g/mol): 244.38 MDL Number: MFCD00042138 InChI Key: XONGRFUEHPBOBB-UHFFFAOYSA-N PubChem CID: 78980 IUPAC Name: 1-chloro-1,1,2-trifluoro-2-iodoethane SMILES: C(C(F)(F)Cl)(F)I