accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFFFI

CAS RN 354-69-8

IUPAC Name: 1,1,1,2,2-pentafluoro-3-iodopropane
Synonyms: 1,1,1,2,2-pentafluoro-3-iodopropane
Molecular Weight (g/mol): 259.95
Molecular Formula: C3H2F5I
InChi Key: HENALDZJQYAUBN-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)CI
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results