accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SO

CAS RN 3541-37-5

IUPAC Name: 1-benzothiophene-2-carbaldehyde
Synonyms: 1-benzothiophene-2-carbaldehyde
Molecular Weight (g/mol): 162.21
Molecular Formula: C9H6OS
InChi Key: NXSVNPSWARVMAY-UHFFFAOYSA-N
SMILES: O=CC1=CC2=CC=CC=C2S1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)