accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3OOOHN

CAS RN 35418-16-7

IUPAC Name: tert-butyl 5-oxopyrrolidine-2-carboxylate
Synonyms: tert-butyl 5-oxopyrrolidine-2-carboxylate
Molecular Weight (g/mol): 185.22
Molecular Formula: C9H15NO3
InChi Key: QXGSPAGZWRTTOT-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)C1CCC(=O)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results