accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3542-36-7

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 418.07
Molecular Formula: C16H36Cl2Sn
InChi Key: DXPJPPSKHFECLR-UHFFFAOYSA-N
SMILES: Cl.Cl.CCCCCCCC[SnH2]CCCCCCCC
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)