accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 35453-19-1

IUPAC Name: 5-amino-2,4,6-triiodobenzene-1,3-dicarboxylate
Synonyms: 5-amino-2,4,6-triiodobenzene-1,3-dicarboxylate
Molecular Weight (g/mol): 556.82
Molecular Formula: C8H2I3NO4
InChi Key: JEZJSNULLBSYHV-UHFFFAOYSA-L
SMILES: NC1=C(I)C(C([O-])=O)=C(I)C(C([O-])=O)=C1I
Molecular Weight (g/mol) 
  • (3)
  • (8)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (2)
  • (6)
  • (3)
  • (8)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (6)