accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 35457-80-8

H3COOOOCH3HOCH3OO(S)O(R)H3C(S)O(S)(R)CH3HO(R)OOCH3(R)H3CO(R)(R)OHNCH3CH3OH3C(E)(Z)

CAS RN 35457-80-8

IUPAC Name: (2R,3R,4R,6S)-6-{[(2R,3S,4R,5R,6S)-4-(dimethylamino)-5-hydroxy-6-{[(11Z,13E)-10-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-7-(2-oxoethyl)-4-(propanoyloxy)-1-oxacyclohexadeca-11,13-dien-6-yl]oxy}-2-methyloxan-3-yl]oxy}-4-hydroxy-2,4-dimethyloxan-3-yl propanoate
Synonyms: midecamycin
Molecular Weight (g/mol): 813.98
Molecular Formula: C41H67NO15
InChi Key: DMUAPQTXSSNEDD-ZZHFWOCQSA-N
SMILES: CCC(=O)O[C@@H]1[C@@H](C)O[C@H](C[C@@]1(C)O)O[C@@H]1[C@@H](C)O[C@@H](OC2C(CC=O)CC(C)C(O)\C=C/C=C/CC(C)OC(=O)CC(OC(=O)CC)C2OC)[C@H](O)[C@H]1N(C)C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)