accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOCH2

CAS RN 35466-83-2

IUPAC Name: methyl prop-2-en-1-yl carbonate
Synonyms: methyl prop-2-en-1-yl carbonate
Molecular Weight (g/mol): 116.12
Molecular Formula: C5H8O3
InChi Key: YHLVIDQQTOMBGN-UHFFFAOYSA-N
SMILES: COC(=O)OCC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results