CAS RN 354811-10-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(5-Phenyl-2-ureido)thiophene-3-carboxamide, TRC
CAS: 354811-10-2 Molecular Formula: C12 H11 N3 O2 S Molecular Weight (g/mol): 261.3 IUPAC Name: 2-(carbamoylamino)-5-phenylthiophene-3-carboxamide SMILES: NC(=O)Nc1sc(cc1C(=O)N)c2ccccc2