accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOFFFFFFFF

CAS RN 355-80-6

IUPAC Name: 2,2,3,3,4,4,5,5-octafluoropentan-1-ol
Synonyms: 2,2,3,3,4,4,5,5-octafluoropentan-1-ol
Molecular Weight (g/mol): 232.07
Molecular Formula: C5H4F8O
InChi Key: JUGSKHLZINSXPQ-UHFFFAOYSA-N
SMILES: OCC(F)(F)C(F)(F)C(F)(F)C(F)F
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)