accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 355423-58-4

IUPAC Name: 6-bromo-7-fluoro-3,4-dihydro-2H-1,4-benzoxazin-3-one
Synonyms: 6-bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one
Molecular Weight (g/mol): 246.04
Molecular Formula: C8H5BrFNO2
InChi Key: AVHJMSMGASUDIT-UHFFFAOYSA-N
SMILES: FC1=C(Br)C=C2NC(=O)COC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results