accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOCH3

CAS RN 35553-92-5

IUPAC Name: ethyl 2-(3-methoxyphenyl)acetate
Synonyms: ethyl 2-(3-methoxyphenyl)acetate
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: XXVVNHCWPHMLEZ-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC=CC(OC)=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)