accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2HOOH2COPOHOHONH2NH2

CAS RN 35556-70-8

IUPAC Name: tris(2-(phosphonooxy)prop-2-enoic acid); cyclohexanamine
Synonyms: cyclohexylamine; tris(phosphoenolpyruvic acid)
Molecular Weight (g/mol): 603.3
Molecular Formula: C15H28NO18P3
InChi Key: WNMMIHPFMKMUPZ-UHFFFAOYSA-N
SMILES: NC1CCCCC1.OC(=O)C(=C)OP(O)(O)=O.OC(=O)C(=C)OP(O)(O)=O.OC(=O)C(=C)OP(O)(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results