accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3556-83-0

IUPAC Name: methyl 3-methoxy-4-methylbenzoate
Synonyms: methyl 3-methoxy-4-methylbenzoate
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: LLEXCSBUSVRBCA-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C)C(OC)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results