accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 35620-71-4

IUPAC Name: 2-phenoxypyridine-3-carboxylate
Synonyms: 2-phenoxypyridine-3-carboxylate
Molecular Weight (g/mol): 214.2
Molecular Formula: C12H8NO3
InChi Key: CQGAXJGXGLVFGJ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CN=C1OC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results