accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3CCClClTi

CAS RN 35625-75-3

IUPAC Name: titanium(4+) bis(2-ethylcyclopenta-1,3-dien-1-ide) dichloride
Synonyms: titanium(4+) bis(2-ethylcyclopenta-1,3-dien-1-ide) dichloride
Molecular Weight (g/mol): 305.07
Molecular Formula: C14H18Cl2Ti
InChi Key: NDYCJOIYEZKDPF-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Ti+4].CCC1=[C-]CC=C1.CCC1=[C-]CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results