accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 35629-70-0

IUPAC Name: 4-methyl-1,3-oxazol-2-amine
Synonyms: 4-methyl-1,3-oxazol-2-amine
Molecular Weight (g/mol): 98.11
Molecular Formula: C4H6N2O
InChi Key: VCZJVXLWQTXSPQ-UHFFFAOYSA-N
SMILES: CC1=COC(N)=N1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results