accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNNBr

CAS RN 356560-80-0

IUPAC Name: 6-bromo-[1,2,4]triazolo[1,5-a]pyridine
Synonyms: 6-bromo-[1,2,4]triazolo[1,5-a]pyridine
Molecular Weight (g/mol): 198.02
Molecular Formula: C6H4BrN3
InChi Key: CXRXKDSDRWLKTK-UHFFFAOYSA-N
SMILES: BrC1=CN2N=CN=C2C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)