accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3567-69-9

NaNaOSOOHNNOSOOO(E)

CAS RN 3567-69-9

IUPAC Name: disodium 4-{2-[(2E)-1-oxo-4-sulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}naphthalene-1-sulfonate
Synonyms: acid red 14
Molecular Weight (g/mol): 502.42
Molecular Formula: C20H12N2Na2O7S2
InChi Key: OJNIRNOPCQQNRY-JZFXEGLSSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=C\C(=N/NC2=C3C=CC=CC3=C(C=C2)S([O-])(=O)=O)C(=O)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Concentration 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results