accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 35691-93-1

H3COOCH3NHNH3C

CAS RN 35691-93-1

IUPAC Name: ethyl 3,5-dimethyl-1H-pyrazole-4-carboxylate
Synonyms: ethyl 3,5-dimethyl-1H-pyrazole-4-carboxylate
Molecular Weight (g/mol): 168.2
Molecular Formula: C8H12N2O2
InChi Key: BCKARVLFIJPHQU-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)NN=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results