accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 35696-77-6

H3CONHSH2NOCH3

CAS RN 35696-77-6

IUPAC Name: (2,4-dimethoxyphenyl)thiourea
Synonyms: 2,4-dimethoxyphenylthiourea
Molecular Weight (g/mol): 212.27
Molecular Formula: C9H12N2O2S
InChi Key: ABBNWHMKEWXTNO-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(NC(N)=S)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results