accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 357-08-4

IUPAC Name: hydrogen (1S,5R,13R,17S)-10,17-dihydroxy-4-(prop-2-en-1-yl)-12-oxa-4-azapentacyclo[9.6.1.0¹,¹³.0⁵,¹⁷.0⁷,¹⁸]octadeca-7,9,11(18)-trien-14-one chloride
Synonyms: hydrogen naloxone chloride
Molecular Weight (g/mol): 363.84
Molecular Formula: C19H22ClNO4
InChi Key: RGPDIGOSVORSAK-STHHAXOLSA-N
SMILES: [H+].[Cl-].OC1=CC=C2C[C@H]3N(CC=C)CC[C@@]45[C@@H](OC1=C24)C(=O)CC[C@@]35O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)