accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OONHOO

CAS RN 35737-10-1

IUPAC Name: 3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoate
Synonyms: 3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoate
Molecular Weight (g/mol): 310.33
Molecular Formula: C18H16NO4
InChi Key: LINBWYYLPWJQHE-UHFFFAOYSA-M
SMILES: [O-]C(=O)CCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results