CAS RN 35793-73-8
CAS RN 35793-73-8
IUPAC Name:
5-(benzyloxy)-2-{[(tert-butoxy)carbonyl]amino}-5-oxopentanoic acid
Synonyms:
5-(benzyloxy)-2-[(tert-butoxycarbonyl)amino]-5-oxopentanoic acid
Molecular Weight (g/mol):
337.37
Molecular Formula:
C17H23NO6
InChi Key:
AJDUMMXHVCMISJ-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC(CCC(=O)OCC1=CC=CC=C1)C(O)=O
1
–
1
of
1
results
1
N-Boc-D-glutamic acid 5-benzyl ester, 98%
CAS: 35793-73-8 Molecular Formula: C17H23NO6 Molecular Weight (g/mol): 337.372 MDL Number: MFCD00038258 InChI Key: AJDUMMXHVCMISJ-CYBMUJFWSA-N Synonym: boc-d-glu obzl-oh,boc-d-glutamic acid 5-benzyl ester,boc-d-glutamic acid 5-benzylester,n-boc-d-glutamic acid 5-benzyl ester,2r-5-benzyloxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid,ambotzbaa1354,boc-d-glutamic acid gamma-benzyl ester,pubchem12172,pubchem14934,boc-d-glu obn-oh PubChem CID: 7010513 IUPAC Name: (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid SMILES: CC(C)(C)OC(=O)NC(CCC(=O)OCC1=CC=CC=C1)C(=O)O