accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SNH3C

CAS RN 3581-91-7

IUPAC Name: 4,5-dimethyl-1,3-thiazole
Synonyms: thiazole, 4,5-dimethyl-
Molecular Weight (g/mol): 113.18
Molecular Formula: C5H7NS
InChi Key: UWSONZCNXUSTKW-UHFFFAOYSA-N
SMILES: CC1=C(C)N=CS1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results