accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C(R)OOOCH3H3C

CAS RN 35825-57-1

IUPAC Name: (1R)-1,6,6-trimethyl-1H,2H,6H,7H,8H,9H,10H,11H-phenanthro[1,2-b]furan-10,11-dione
Synonyms: cryptotanshinone
Molecular Weight (g/mol): 296.37
Molecular Formula: C19H20O3
InChi Key: GVKKJJOMQCNPGB-JTQLQIEISA-N
SMILES: C[C@H]1COC2=C1C(=O)C(=O)C1=C3CCCC(C)(C)C3=CC=C21
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)