accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3586-14-9

IUPAC Name: 1-methyl-3-phenoxybenzene
Synonyms: benzene, 1-methyl-3-phenoxy-
Molecular Weight (g/mol): 184.24
Molecular Formula: C13H12O
InChi Key: UDONPJKEOAWFGI-UHFFFAOYSA-N
SMILES: CC1=CC=CC(OC2=CC=CC=C2)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results