accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOOHO(E)(E)

CAS RN 3588-17-8

IUPAC Name: (2E,4E)-hexa-2,4-dienedioic acid
Synonyms: trans, trans-muconic acid trans,trans-muconic acid trans-muconic acid trans
Molecular Weight (g/mol): 142.11
Molecular Formula: C6H6O4
InChi Key: TXXHDPDFNKHHGW-ZPUQHVIOSA-N
SMILES: OC(=O)\C=C\C=C\C(O)=O
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)