accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOO

CAS RN 35888-99-4

IUPAC Name: 3-{[1,1'-biphenyl]-4-yl}propanoic acid
Synonyms: 3-{[1,1'-biphenyl]-4-yl}propanoic acid
Molecular Weight (g/mol): 226.28
Molecular Formula: C15H14O2
InChi Key: MVFHRQWYCXYYMU-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC=C(C=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results