accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNH3CI

CAS RN 35935-34-3

IUPAC Name: 1-ethyl-3-methyl-1H-imidazol-3-ium iodide
Synonyms: 1-ethyl-3-methylimidazolium iodide
Molecular Weight (g/mol): 238.07
Molecular Formula: C6H11IN2
InChi Key: IKQCDTXBZKMPBB-UHFFFAOYSA-M
SMILES: [I-].CCN1C=C[N+](C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results