accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 35948-25-5

IUPAC Name: 9-oxo-8-oxa-9λ⁵-phosphatricyclo[8.4.0.0²,⁷]tetradeca-1(14),2,4,6,10,12-hexaen-9-ylium
Synonyms: 9-oxo-8-oxa-9λ⁵-phosphatricyclo[8.4.0.0²,⁷]tetradeca-1(14),2,4,6,10,12-hexaen-9-ylium
Molecular Weight (g/mol): 215.17
Molecular Formula: C12H8O2P
InChi Key: DWSWCPPGLRSPIT-UHFFFAOYNA-N
SMILES: O=[P+]1OC2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)