accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClON

CAS RN 3598-13-8

IUPAC Name: 2-(4-chlorophenoxy)acetonitrile
Synonyms: 2-(4-chlorophenoxy)acetonitrile
Molecular Weight (g/mol): 167.59
Molecular Formula: C8H6ClNO
InChi Key: YUGDKEWUYZXXRU-UHFFFAOYSA-N
SMILES: ClC1=CC=C(OCC#N)C=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)