CAS RN 36062-04-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tetrahydro Curcumin, TRC
CAS: 36062-04-1 Molecular Formula: C21 H24 O6 Molecular Weight (g/mol): 372.41 Synonym: 3,5-Heptanedione, 1,7-bis(4-hydroxy-3-methoxyphenyl)-,1,7-Bis(4-hydroxy-3-methoxyphenyl)-3,5-heptanedione,1,7-Bis(4-hydroxy-3-methoxyphenyl)heptane-3,5-dione,AS-KTC 004,HZIV 81-2,NSC 687845,Sabiwhite,Tetrahydrocurcumin,Tetrahydrodiferuloylmethane IUPAC Name: 1,7-bis(4-hydroxy-3-methoxyphenyl)heptane-3,5-dione SMILES: COc1cc(CCC(=O)CC(=O)CCc2ccc(O)c(OC)c2)ccc1O