accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 36062-19-8

HClNH2HNNH2NH2N(E)

CAS RN 36062-19-8

IUPAC Name: hydrogen (E)-N,N''-diaminoguanidine chloride
Synonyms: hydrogen (E)-N,N''-diaminoguanidine chloride
Molecular Weight (g/mol): 125.56
Molecular Formula: CH8ClN5
InChi Key: HAZRIBSLCUYMQP-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NN\C(N)=N\N
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)